Product Details
- SNP ID
-
rs17851676
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:98282365 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCGCCCACAGGGATACCCACCCAC[C/T]ATCCCAGGGTCTACAACATCCACAG
- Phenotype
-
MIM: 611877
MIM: 615628
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BAIAP2L1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147351220] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BAIAP2L1
- Gene Name
- BAI1 associated protein 2 like 1
There are no transcripts associated with this gene.
- Gene
- BRI3
- Gene Name
- brain protein I3
View Full Product Details