Product Details

SNP ID
rs16863317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151326671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTATAAAGTTCAAGAGATGGATA[A/G]GTAGCTAGGAGGTAAGGCCAGAAAG
Phenotype
MIM: 606379 MIM: 611318 MIM: 606380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR87 PubMed Links

Gene Details

Gene
GPR87
Gene Name
G protein-coupled receptor 87
There are no transcripts associated with this gene.

Gene
MED12L
Gene Name
mediator complex subunit 12 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053002.5 2406 Intron NP_443728.3
XM_006713487.3 2406 Intron XP_006713550.1
XM_011512390.2 2406 Intron XP_011510692.1
XM_011512394.2 2406 Intron XP_011510696.1
XM_011512399.2 2406 Intron XP_011510701.1
XM_017005676.1 2406 Intron XP_016861165.1
XM_017005677.1 2406 Intron XP_016861166.1
XM_017005678.1 2406 Intron XP_016861167.1
XM_017005679.1 2406 Intron XP_016861168.1
XM_017005680.1 2406 Intron XP_016861169.1
XM_017005681.1 2406 Intron XP_016861170.1
Gene
P2RY13
Gene Name
purinergic receptor P2Y13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176894.2 2406 UTR 3 NP_795713.2
XM_006713664.1 2406 UTR 3 XP_006713727.1

View Full Product Details