Product Details

SNP ID
rs8014252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:70452982 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTAAGGTAGAGAAGAGGTGGATA[C/T]AGGCAGATTCAGATGGTGAAAGGCC
Phenotype
MIM: 603713
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAM21 PubMed Links

Gene Details

Gene
ADAM21
Gene Name
ADAM metallopeptidase domain 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003813.3 Intron NP_003804.2

View Full Product Details