Product Details
- SNP ID
-
rs17129325
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:93908756 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCATATGTCTGTCCATCAAACATTG[C/T]TGAGCCCTGAGCTACTGGTGCCTTA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM181A-AS1
PubMed Links
Gene Details
- Gene
- FAM181A-AS1
- Gene Name
- FAM181A antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details