Product Details

SNP ID
rs17859841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:55480235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAGATCCGGAGAGGGACCTTTA[A/G]ACAAACAGCTCGAGGGGGCCATTTG
Phenotype
MIM: 120360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMP2 PubMed Links

Gene Details

Gene
MMP2
Gene Name
matrix metallopeptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127891.2 Intron NP_001121363.1
NM_001302508.1 Intron NP_001289437.1
NM_001302509.1 Intron NP_001289438.1
NM_001302510.1 Intron NP_001289439.1
NM_004530.5 Intron NP_004521.1

View Full Product Details