Product Details
- SNP ID
-
rs16963745
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:61646672 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTCCCATCCATCTGTTTTGTTCT[A/G]TCGTATCAAGTTTTTTTTATTTCCA
- Phenotype
-
MIM: 603008
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CDH8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142486944] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDH8
- Gene Name
- cadherin 8
View Full Product Details