Product Details

SNP ID
rs17841889
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35243355 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGGCCTCTGTTTTTGAGAATGCA[C/G]AATTAATTGCACTTGACAAACTTGC
Phenotype
MIM: 609601
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ZNF397 PubMed Links

Gene Details

Gene
ZNF397
Gene Name
zinc finger protein 397
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135178.2 Intron NP_001128650.1
NM_032347.2 Intron NP_115723.1
XM_006722558.2 Intron XP_006722621.1
XM_011526229.2 Intron XP_011524531.1
XM_011526230.2 Intron XP_011524532.1
XM_011526231.2 Intron XP_011524533.1
XM_017026042.1 Intron XP_016881531.1
Gene
ZSCAN30
Gene Name
zinc finger and SCAN domain containing 30
There are no transcripts associated with this gene.

View Full Product Details