Product Details

SNP ID
rs17060532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:174492145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAACGAAAGAATGAGGCATATTAC[C/T]ACTTCATTTTAAGTTATTTTCTGAA
Phenotype
MIM: 601688
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HPGD PubMed Links

Gene Details

Gene
HPGD
Gene Name
hydroxyprostaglandin dehydrogenase 15-(NAD)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000860.5 Intron NP_000851.2
NM_001145816.2 Intron NP_001139288.1
NM_001256301.1 Intron NP_001243230.1
NM_001256305.1 Intron NP_001243234.1
NM_001256306.1 Intron NP_001243235.1
NM_001256307.1 Intron NP_001243236.1
XM_011531907.1 Intron XP_011530209.1

View Full Product Details