Product Details

SNP ID
hCV33825557
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.2:108289027 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
AAGTCCTTTCCGTGCCCACTGACCC[-/C]TTGAGTGGGCCTTTGAGCTGCTGAC
Phenotype
MIM: 602385
Polymorphism
-/C, Insertion/deletion
Allele Nomenclature
Literature Links
SULT1C2 PubMed Links

Gene Details

Gene
SULT1C2
Gene Name
sulfotransferase family 1C member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001056.3 389 UTR 5 NP_001047.1
NM_176825.2 389 UTR 5 NP_789795.1

View Full Product Details