Product Details

SNP ID
rs17123627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49241295 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATGAGAATGGGGCCAAGATTAT[A/C]ATGTTCATTTTGCACCCCCAGTGCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TUBA1C PubMed Links

Gene Details

Gene
TUBA1C
Gene Name
tubulin alpha 1c
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303114.1 Intron NP_001290043.1
NM_001303115.1 Intron NP_001290044.1
NM_001303116.1 Intron NP_001290045.1
NM_001303117.1 Intron NP_001290046.1
NM_032704.4 Intron NP_116093.1

View Full Product Details