Product Details
- SNP ID
-
rs35070488
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:39110883 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAATTCAGGAGCTTGAGTACAGC[C/T]TGAAGGAAATACGCAAGACAGTCAG
- Phenotype
-
MIM: 606867
MIM: 611430
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GORASP1
PubMed Links
Gene Details
- Gene
- GORASP1
- Gene Name
- golgi reassembly stacking protein 1
There are no transcripts associated with this gene.
- Gene
- TTC21A
- Gene Name
- tetratricopeptide repeat domain 21A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001105513.2 |
428 |
Silent Mutation |
CTG,TTG |
L101L |
NP_001098983.2 |
NM_145755.2 |
428 |
Silent Mutation |
CTG,TTG |
L101L |
NP_665698.2 |
XM_005264921.4 |
428 |
Silent Mutation |
CTG,TTG |
L101L |
XP_005264978.1 |
XM_005264922.4 |
428 |
Silent Mutation |
CTG,TTG |
L101L |
XP_005264979.1 |
XM_005264923.3 |
428 |
Silent Mutation |
CTG,TTG |
L101L |
XP_005264980.1 |
XM_005264924.4 |
428 |
Silent Mutation |
CTG,TTG |
L101L |
XP_005264981.1 |
XM_005264925.4 |
428 |
Silent Mutation |
CTG,TTG |
L101L |
XP_005264982.1 |
XM_005264926.4 |
428 |
Silent Mutation |
CTG,TTG |
L101L |
XP_005264983.1 |
XM_005264927.4 |
428 |
Intron |
|
|
XP_005264984.1 |
XM_006713011.3 |
428 |
Silent Mutation |
CTG,TTG |
L101L |
XP_006713074.1 |
XM_011533447.2 |
428 |
Silent Mutation |
CTG,TTG |
L101L |
XP_011531749.1 |
XM_011533449.1 |
428 |
Silent Mutation |
CTG,TTG |
L101L |
XP_011531751.1 |
XM_017005843.1 |
428 |
Silent Mutation |
CTG,TTG |
L101L |
XP_016861332.1 |
View Full Product Details