Product Details

SNP ID
rs17816402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:12382636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATTTTTGTTCTTGAGCAATCTT[A/T]CACCCAAGATTCTGTGGTTCTGCGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM66A PubMed Links

Gene Details

Gene
FAM66A
Gene Name
family with sequence similarity 66 member A
There are no transcripts associated with this gene.

Gene
LOC649352
Gene Name
ubiquitin carboxyl-terminal hydrolase 17-like protein 2-like
There are no transcripts associated with this gene.

View Full Product Details