Product Details
- SNP ID
-
rs17462807
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:13967599 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTTGTTTTTACATACAGATGTTA[C/T]ATGACCATGCCTGTAAAATTATATG
- Phenotype
-
MIM: 604989
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SPON1
PubMed Links
Gene Details
- Gene
- SPON1
- Gene Name
- spondin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006108.3 |
|
Intron |
|
|
NP_006099.2 |
View Full Product Details