Product Details

SNP ID
rs34967265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:929741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCATGCGGGCTGCAGCTGCGGG[C/T]CTGGGACACCCAGCCAGCCCCGGCG
Phenotype
MIM: 603265 MIM: 604161
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARID3A PubMed Links

Gene Details

Gene
ARID3A
Gene Name
AT-rich interaction domain 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005224.2 668 Silent Mutation GGC,GGT G71G NP_005215.1
XM_005259513.4 668 Silent Mutation GGC,GGT G71G XP_005259570.1
XM_005259514.4 668 Silent Mutation GGC,GGT G71G XP_005259571.1
XM_017026445.1 668 Silent Mutation GGC,GGT G71G XP_016881934.1
XM_017026446.1 668 Intron XP_016881935.1
XM_017026447.1 668 Intron XP_016881936.1
Gene
KISS1R
Gene Name
KISS1 receptor
There are no transcripts associated with this gene.

View Full Product Details