Product Details

SNP ID
rs17036350
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:11111169 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCCTTTCTACCCATCAGATTCCA[C/T]CCATCCAATAAGGATAAATTACCCT
Phenotype
MIM: 601231
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105376736 PubMed Links

Gene Details

Gene
LOC105376736
Gene Name
uncharacterized LOC105376736
There are no transcripts associated with this gene.

Gene
MTOR
Gene Name
mechanistic target of rapamycin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004958.3 Intron NP_004949.1
XM_005263438.2 Intron XP_005263495.1
XM_011541166.2 Intron XP_011539468.1
XM_017000900.1 Intron XP_016856389.1
XM_017000901.1 Intron XP_016856390.1
XM_017000902.1 Intron XP_016856391.1

View Full Product Details