Product Details

SNP ID
rs17579825
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55510141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCTGCACTACACAACCATTATGC[A/G]ACAAGGGATCTGCCACCTTCTGGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4C1P PubMed Links

Gene Details

Gene
OR4C1P
Gene Name
olfactory receptor family 4 subfamily C member 1 pseudogene
There are no transcripts associated with this gene.

View Full Product Details