Product Details
- SNP ID
-
rs17148883
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:55914096 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACCCACAGAAGCATAGGCGGAAGG[C/G]CAGTGTCATATGTATCAAAGCATCT
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR5W2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17596519,rs61749302] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR5W2
- Gene Name
- olfactory receptor family 5 subfamily W member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001960.1 |
487 |
Missense Mutation |
CCC,GCC |
P163A |
NP_001001960.1 |
View Full Product Details