Product Details

SNP ID
rs16842121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:8366442 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATCAAACAGATGTTCCTCCTTAA[C/G]TCTTAAACCACGACATTCAGATGAT
Phenotype
MIM: 603402
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACOX3 PubMed Links

Gene Details

Gene
ACOX3
Gene Name
acyl-CoA oxidase 3, pristanoyl
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101667.1 2646 UTR 3 NP_001095137.1
NM_003501.2 2646 UTR 3 NP_003492.2
XM_005248011.4 2646 UTR 3 XP_005248068.1
XM_005248013.3 2646 UTR 3 XP_005248070.1
XM_011513565.2 2646 UTR 3 XP_011511867.1
XM_011513566.1 2646 UTR 3 XP_011511868.1

View Full Product Details