Product Details
- SNP ID
-
rs16967444
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:57186762 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTCTTGGGGGTCGCCCTTCTGAG[C/G]AGGCTGCGGCTAACAGGGCCCAGGT
- Phenotype
-
MIM: 616585
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM192A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77056538] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM192A
- Gene Name
- family with sequence similarity 192 member A
- Gene
- RSPRY1
- Gene Name
- ring finger and SPRY domain containing 1
View Full Product Details