Product Details

SNP ID
rs17849335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58708181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGCACCAGGGACATTACTTGGTG[A/G]CCTGGTGAATGGCATGGGCAAGGTA
Phenotype
MIM: 138150
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOT2 PubMed Links

Gene Details

Gene
GOT2
Gene Name
glutamic-oxaloacetic transaminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286220.1 Intron NP_001273149.1
NM_002080.3 Intron NP_002071.2

View Full Product Details