Product Details

SNP ID
rs16957128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67044082 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTCCTCTTAGGACTTATTTCCA[A/T]TTACTTTTATTTCAAATAAATGCAA
Phenotype
MIM: 121360
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CBFB PubMed Links

Gene Details

Gene
CBFB
Gene Name
core-binding factor, beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001755.2 Intron NP_001746.1
NM_022845.2 Intron NP_074036.1
XM_005256212.3 Intron XP_005256269.1
XM_006721321.2 Intron XP_006721384.1
XM_017023812.1 Intron XP_016879301.1
XM_017023813.1 Intron XP_016879302.1

View Full Product Details