Product Details

SNP ID
rs16979414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9294108 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACGTATTTTTCCCCATGAATAGCA[C/T]GACATGATGCGATTGGACTGTGTAA
Phenotype
MIM: 609571
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF699 PubMed Links

Gene Details

Gene
ZNF699
Gene Name
zinc finger protein 699
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198535.2 3666 UTR 3 NP_940937.1

View Full Product Details