Product Details

SNP ID
rs17855476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82322337 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGCTCCCGCTCTGAGGGCTGCGA[A/C]CTTCATCTGGGGTTCTAGGAGGAAG
Phenotype
MIM: 186820 MIM: 602602
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CD7 PubMed Links

Gene Details

Gene
CD7
Gene Name
CD7 molecule
There are no transcripts associated with this gene.

Gene
SECTM1
Gene Name
secreted and transmembrane 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003004.2 1000 Missense Mutation GTC,TTC V191F NP_002995.1
XM_005256392.3 1000 Missense Mutation GTC,TTC V191F XP_005256449.1
XM_011523588.2 1000 Missense Mutation GTC,TTC V191F XP_011521890.1

View Full Product Details