Product Details

SNP ID
rs35198568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:58870898 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGACTTTAACCCCAACATATTCT[C/T]TAAGAAGAATCTGCTTTTTGAATAG
Phenotype
MIM: 608870
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRIG3 PubMed Links

Gene Details

Gene
LRIG3
Gene Name
leucine rich repeats and immunoglobulin like domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136051.2 5009 Intron NP_001129523.1
NM_153377.4 5009 Intron NP_700356.2
XM_011537893.2 5009 UTR 3 XP_011536195.1
XM_017018790.1 5009 Intron XP_016874279.1

View Full Product Details