Product Details

SNP ID
rs17729501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:668618 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCAGTATTCCCTTATTGAACACC[C/T]ATTTTTAAATATCTGGGTCTCTAAC
Phenotype
MIM: 613220
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM18 PubMed Links

Gene Details

Gene
TMEM18
Gene Name
transmembrane protein 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152834.2 1479 UTR 3 NP_690047.2

View Full Product Details