Product Details

SNP ID
rs17700084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:46683274 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTTAAATCAAGTGGTTGGTCTA[C/T]AAATATTCATTTTATTGTTTTGCTT
Phenotype
MIM: 607162
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ST8SIA5 PubMed Links

Gene Details

Gene
ST8SIA5
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307986.1 Intron NP_001294915.1
NM_001307987.1 Intron NP_001294916.1
NM_013305.5 Intron NP_037437.2
XM_017025715.1 Intron XP_016881204.1
XM_017025716.1 Intron XP_016881205.1

View Full Product Details