Product Details

SNP ID
rs17057105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:96899229 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCAGTGATCTCAGATAGCATTAA[C/T]TGGGAAGTTATCCTGAAGGTGGCAG
Phenotype
MIM: 611776
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL32 PubMed Links

Gene Details

Gene
KLHL32
Gene Name
kelch like family member 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286250.1 1147 Intron NP_001273179.1
NM_001286251.1 1147 Intron NP_001273180.1
NM_001286252.1 1147 Intron NP_001273181.1
NM_001286254.2 1147 Intron NP_001273183.1
NM_001323252.1 1147 Intron NP_001310181.1
NM_001323253.1 1147 Intron NP_001310182.1
NM_001323254.1 1147 Intron NP_001310183.1
NM_001323255.1 1147 Intron NP_001310184.1
NM_001323256.1 1147 Intron NP_001310185.1
NM_001323257.1 1147 Intron NP_001310186.1
NM_001323258.1 1147 UTR 5 NP_001310187.1
NM_001323259.1 1147 Intron NP_001310188.1
NM_001323260.1 1147 Intron NP_001310189.1
NM_001323261.1 1147 Intron NP_001310190.1
NM_001323262.1 1147 Intron NP_001310191.1
NM_001323263.1 1147 Intron NP_001310192.1
NM_001323264.1 1147 Intron NP_001310193.1
NM_052904.3 1147 Intron NP_443136.2
XM_005266813.4 1147 Intron XP_005266870.1
XM_017010228.1 1147 Intron XP_016865717.1
XM_017010229.1 1147 Intron XP_016865718.1
Gene
NDUFAF4
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 4
There are no transcripts associated with this gene.

View Full Product Details