Product Details
- SNP ID
-
rs17289116
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:32457531 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACAATAATCTGAGGCAAAGAACAG[A/G]TGGGTCTCACTTATTTTACAATTGG
- Phenotype
-
MIM: 100880
MIM: 609631
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACO1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75836526] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACO1
- Gene Name
- aconitase 1
There are no transcripts associated with this gene.
- Gene
- DDX58
- Gene Name
- DEXD/H-box helicase 58
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014314.3 |
|
Intron |
|
|
NP_055129.2 |
- Gene
- LOC101060445
- Gene Name
- uncharacterized LOC101060445
There are no transcripts associated with this gene.
View Full Product Details