Product Details

SNP ID
rs16989988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:32190707 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTATAATGCTTTTACGAAGTAGAG[C/T]GTTCCTAAGTCTACCCACCCAAGTA
Phenotype
MIM: 605969
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RFPL2 PubMed Links

Gene Details

Gene
RFPL2
Gene Name
ret finger protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098527.2 2252 UTR 3 NP_001091997.2
NM_001159545.1 2252 UTR 3 NP_001153017.1
NM_001159546.1 2252 Intron NP_001153018.1
NM_006605.3 2252 Intron NP_006596.2
XM_011529826.1 2252 UTR 3 XP_011528128.1
XM_011529827.2 2252 UTR 3 XP_011528129.1
XM_011529828.1 2252 UTR 3 XP_011528130.1
XM_011529830.2 2252 UTR 3 XP_011528132.1
XM_011529832.1 2252 UTR 3 XP_011528134.1
XM_017028535.1 2252 UTR 3 XP_016884024.1
XM_017028536.1 2252 UTR 3 XP_016884025.1

View Full Product Details