Product Details

SNP ID
rs7286723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42380962 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTGAAGGGAACCCCAGCCACACC[A/G]GGTTCAGCGCAGAACCAGCACATGT
Phenotype
MIM: 608740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NFAM1 PubMed Links

Gene Details

Gene
NFAM1
Gene Name
NFAT activating protein with ITAM motif 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318323.1 5677 UTR 3 NP_001305252.1
NM_145912.6 5677 UTR 3 NP_666017.1
XM_005261374.4 5677 UTR 3 XP_005261431.1

View Full Product Details