Product Details

SNP ID
rs35926160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:123149723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGAGCGGCCGCGGGACGGCGGCG[A/C]TGAGCGGCAGGCCGGCGCGGCCAAG
Phenotype
MIM: 603719 MIM: 600647
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BUB3 PubMed Links

Gene Details

Gene
BUB3
Gene Name
BUB3, mitotic checkpoint protein
There are no transcripts associated with this gene.

Gene
HMX2
Gene Name
H6 family homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005519.1 679 Missense Mutation GAT,GCT D141A NP_005510.1
XM_005269743.3 679 Missense Mutation GAT,GCT D141A XP_005269800.1

View Full Product Details