Product Details

SNP ID
rs17257382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24813134 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAATTGTATTTACTGGCATCTCA[C/T]GAAGTATTAAAAATCAACAGCTTTT
Phenotype
MIM: 607958
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STXBP6 PubMed Links

Gene Details

Gene
STXBP6
Gene Name
syntaxin binding protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304476.1 Intron NP_001291405.1
NM_001304477.1 Intron NP_001291406.1
NM_014178.7 Intron NP_054897.4
XM_006720121.3 Intron XP_006720184.1
XM_011536669.2 Intron XP_011534971.1
XM_017021231.1 Intron XP_016876720.1
XM_017021232.1 Intron XP_016876721.1
XM_017021233.1 Intron XP_016876722.1
XM_017021234.1 Intron XP_016876723.1
XM_017021235.1 Intron XP_016876724.1
XM_017021236.1 Intron XP_016876725.1
XM_017021237.1 Intron XP_016876726.1
XM_017021238.1 Intron XP_016876727.1
XM_017021239.1 Intron XP_016876728.1
XM_017021240.1 Intron XP_016876729.1
XM_017021241.1 Intron XP_016876730.1
XM_017021242.1 Intron XP_016876731.1

View Full Product Details