Product Details

SNP ID
rs17292947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:72743330 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTACTATTTGACCATTGTCTCCAT[C/T]CTTCAAGACCTGCTTAGAAGGTCAG
Phenotype
MIM: 607973
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NETO1 PubMed Links

Gene Details

Gene
NETO1
Gene Name
neuropilin and tolloid like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201465.1 6254 Intron NP_001188394.1
NM_138966.3 6254 Intron NP_620416.1
NM_138999.1 6254 Intron NP_620552.1
XM_005266774.4 6254 UTR 3 XP_005266831.1
XM_017026017.1 6254 Intron XP_016881506.1
XM_017026018.1 6254 Intron XP_016881507.1
XM_017026019.1 6254 Intron XP_016881508.1
XM_017026020.1 6254 UTR 3 XP_016881509.1
XM_017026021.1 6254 Intron XP_016881510.1
XM_017026022.1 6254 Intron XP_016881511.1
XM_017026023.1 6254 Intron XP_016881512.1
XM_017026024.1 6254 Intron XP_016881513.1

View Full Product Details