Product Details

SNP ID
rs17851549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4652222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTCGACCGGCGCGTGCTGGCCG[C/T]CGGGCTGCTCGAGTGCCGCGACCTG
Phenotype
MIM: 606746 MIM: 615869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYDGF PubMed Links

Gene Details

Gene
MYDGF
Gene Name
myeloid derived growth factor
There are no transcripts associated with this gene.

Gene
TNFAIP8L1
Gene Name
TNF alpha induced protein 8 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167942.1 1186 Missense Mutation GCC,GTC A118V NP_001161414.1
NM_152362.2 1186 Missense Mutation GCC,GTC A118V NP_689575.2
XM_005259487.3 1186 Missense Mutation GCC,GTC A118V XP_005259544.1
XM_011527680.2 1186 Missense Mutation GCC,GTC A118V XP_011525982.1

View Full Product Details