Product Details

SNP ID
rs16963485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50242506 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTGTTGCAACTGAGGGAGCGCAC[C/T]GTCTTCTTCTTAGTCTGCACAGACA
Phenotype
MIM: 142704 MIM: 603247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HDC PubMed Links

Gene Details

Gene
HDC
Gene Name
histidine decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306146.1 1954 Silent Mutation ACA,ACG T548T NP_001293075.1
NM_002112.3 1954 Silent Mutation ACA,ACG T581T NP_002103.2
XM_017022094.1 1954 Silent Mutation ACA,ACG T616T XP_016877583.1
XM_017022095.1 1954 Silent Mutation ACA,ACG T583T XP_016877584.1
XM_017022096.1 1954 Silent Mutation ACA,ACG T540T XP_016877585.1
XM_017022097.1 1954 Silent Mutation ACA,ACG T537T XP_016877586.1
XM_017022098.1 1954 Silent Mutation ACA,ACG T472T XP_016877587.1
XM_017022099.1 1954 Intron XP_016877588.1
Gene
SLC27A2
Gene Name
solute carrier family 27 member 2
There are no transcripts associated with this gene.

View Full Product Details