Product Details

SNP ID
rs16940768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:58771307 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCGAGGCCGCGCCAGGGCGCTGT[A/T]GCTGCCAATACAGCTGTCATGGCGT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM63B PubMed Links

Gene Details

Gene
FAM63B
Gene Name
family with sequence similarity 63 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040450.2 114 UTR 5 NP_001035540.1
NM_001040453.2 114 Intron NP_001035543.1
XM_011521687.1 114 UTR 5 XP_011519989.1
XM_011521688.1 114 UTR 5 XP_011519990.1
XM_011521689.1 114 UTR 5 XP_011519991.1
XM_011521690.1 114 UTR 5 XP_011519992.1
XM_017022332.1 114 UTR 5 XP_016877821.1
XM_017022333.1 114 UTR 5 XP_016877822.1
XM_017022334.1 114 UTR 5 XP_016877823.1
XM_017022335.1 114 UTR 5 XP_016877824.1
Gene
LOC101928725
Gene Name
uncharacterized LOC101928725
There are no transcripts associated with this gene.

View Full Product Details