Product Details

SNP ID
rs17091195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94645251 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACAACCTGCTTTGTTAAAACACTG[A/G]GGATCCCAAAGGAAATAAGGAAAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINA13P PubMed Links

Gene Details

Gene
SERPINA13P
Gene Name
serpin family A member 13, pseudogene
There are no transcripts associated with this gene.

View Full Product Details