Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014877.3 | 13260 | UTR 3 | NP_055692.2 | ||
XM_005257888.4 | 13260 | UTR 3 | XP_005257945.1 | ||
XM_005257889.4 | 13260 | UTR 3 | XP_005257946.1 | ||
XM_006722214.3 | 13260 | UTR 3 | XP_006722277.1 | ||
XM_006722215.3 | 13260 | UTR 3 | XP_006722278.1 | ||
XM_006722216.3 | 13260 | UTR 3 | XP_006722279.1 | ||
XM_011525544.2 | 13260 | UTR 3 | XP_011523846.1 | ||
XM_017025477.1 | 13260 | UTR 3 | XP_016880966.1 | ||
XM_017025478.1 | 13260 | UTR 3 | XP_016880967.1 |