Product Details

SNP ID
rs16973554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:69063256 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCCTTTTGCTTCATTTATTTTGC[A/G]GCAGTATTCTCCAGTGCCAGAAACC
Phenotype
MIM: 612504 MIM: 612507
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCA6 PubMed Links

Gene Details

Gene
ABCA6
Gene Name
ATP binding cassette subfamily A member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080284.2 Intron NP_525023.2
XM_011524574.2 Intron XP_011522876.1
XM_017024404.1 Intron XP_016879893.1
XM_017024405.1 Intron XP_016879894.1
XM_017024406.1 Intron XP_016879895.1
XM_017024407.1 Intron XP_016879896.1
XM_017024408.1 Intron XP_016879897.1
Gene
ABCA9
Gene Name
ATP binding cassette subfamily A member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080283.3 Intron NP_525022.2
XM_017024011.1 Intron XP_016879500.1
XM_017024012.1 Intron XP_016879501.1
XM_017024013.1 Intron XP_016879502.1
XM_017024014.1 Intron XP_016879503.1
XM_017024015.1 Intron XP_016879504.1

View Full Product Details