Product Details

SNP ID
rs16858781
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:167231083 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCAGAAAGGCCCTATACTGGAAAG[A/G]CAGACTGAGAATTAGGAGACTTGGA
Phenotype
MIM: 164175
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POU2F1 PubMed Links

Gene Details

Gene
POU2F1
Gene Name
POU class 2 homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198783.1 Intron NP_001185712.1
NM_001198786.1 Intron NP_001185715.1
NM_002697.3 Intron NP_002688.3
XM_011509653.2 Intron XP_011507955.1
XM_011509654.2 Intron XP_011507956.1
XM_011509655.1 Intron XP_011507957.1
XM_017001508.1 Intron XP_016856997.1

View Full Product Details