Product Details
- SNP ID
-
rs16861337
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
9
- Location
-
Chr.1:168697755 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTCTCTTTTAATCCTCACAACGA[C/T]TCATTGTTTAGGTGAAAAACCAGGC
- Phenotype
-
MIM: 125597
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DPT
PubMed Links
Gene Details
- Gene
- DPT
- Gene Name
- dermatopontin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001937.4 |
|
Intron |
|
|
NP_001928.2 |
View Full Product Details