Product Details

SNP ID
rs16898570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:100551401 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTAAGTGAGCCCCAGCCTTCTC[C/T]ATGGTCATGAAGATGCCAGTGGACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD46 PubMed Links

Gene Details

Gene
ANKRD46
Gene Name
ankyrin repeat domain 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270377.1 Intron NP_001257306.1
NM_001270378.1 Intron NP_001257307.1
NM_001270379.1 Intron NP_001257308.1
NM_198401.3 Intron NP_940683.1
XM_006716509.1 Intron XP_006716572.1
XM_017013106.1 Intron XP_016868595.1
Gene
GAPDHP62
Gene Name
glyceraldehyde 3 phosphate dehydrogenase pseudogene 62
There are no transcripts associated with this gene.

View Full Product Details