Product Details

SNP ID
rs16892524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:119417239 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGACAAGAATCTAAGCAAAGGGG[C/G]AGCCACGCAGAGTTAAGGGCCAGTG
Phenotype
MIM: 164958
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NOV PubMed Links

Gene Details

Gene
NOV
Gene Name
nephroblastoma overexpressed
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002514.3 Intron NP_002505.1

View Full Product Details