Product Details

SNP ID
rs74315368
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:17022648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCAGTTCATGATGGTGTGGCAG[T/C]GGTATAGAGAGAATGGGTCCTGCAG
Phenotype
MIM: 185470
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SDHB PubMed Links

Gene Details

Gene
SDHB
Gene Name
succinate dehydrogenase complex iron sulfur subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003000.2 876 Missense Mutation CAC,CGC H242R NP_002991.2

View Full Product Details