Product Details

SNP ID
rs16939670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:85778878 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTCAATTCCCTAAGGTTTGGTTA[C/G]AAAACTTGACCAAATGTTCTAAGTA
Phenotype
MIM: 604886
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EMC8 PubMed Links

Gene Details

Gene
EMC8
Gene Name
ER membrane protein complex subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142288.1 1401 UTR 3 NP_001135760.1
NM_006067.4 1401 UTR 3 NP_006058.1
XM_011522813.2 1401 Intron XP_011521115.1
XM_017022867.1 1401 UTR 3 XP_016878356.1

View Full Product Details