Product Details

SNP ID
rs17084624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:95393935 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGACAGTTTTTATATATAATAACA[A/T]CCTAGCTTATAGGAATGAGGATAAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM81B PubMed Links

Gene Details

Gene
FAM81B
Gene Name
family with sequence similarity 81 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152548.2 Intron NP_689761.2
XM_005271905.2 Intron XP_005271962.2
XM_005271906.4 Intron XP_005271963.2
XM_005271907.3 Intron XP_005271964.1
XM_011543207.1 Intron XP_011541509.1
XM_011543208.1 Intron XP_011541510.1
XM_011543209.1 Intron XP_011541511.1
XM_011543210.2 Intron XP_011541512.1
XM_017009118.1 Intron XP_016864607.1

View Full Product Details