Product Details

SNP ID
rs17120527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:64622520 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAAAGGAATCGGCATTGGCATCT[A/G]TTAGGTTGGTGCAAAAGTAATTGCG
Phenotype
MIM: 607019
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR548C PubMed Links

Gene Details

Gene
MIR548C
Gene Name
microRNA 548c
There are no transcripts associated with this gene.

Gene
MIR548Z
Gene Name
microRNA 548z
There are no transcripts associated with this gene.

Gene
RASSF3
Gene Name
Ras association domain family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178169.3 Intron NP_835463.1
XM_006719345.3 Intron XP_006719408.1
XM_011538195.2 Intron XP_011536497.1
XM_017019182.1 Intron XP_016874671.1

View Full Product Details