Product Details

SNP ID
rs16896400
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43297569 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTACCCCTCCAAAAGGTAATCTT[A/G]GGCATTGCAGGTACCTTGGCAACAT
Phenotype
MIM: 604995
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CRIP3 PubMed Links

Gene Details

Gene
CRIP3
Gene Name
cysteine rich protein 3
There are no transcripts associated with this gene.

Gene
SLC22A7
Gene Name
solute carrier family 22 member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006672.3 Intron NP_006663.2
NM_153320.2 Intron NP_696961.2
XM_006714970.3 Intron XP_006715033.1
XM_006714971.3 Intron XP_006715034.1
XM_011514256.2 Intron XP_011512558.1
XM_011514257.2 Intron XP_011512559.1
XM_011514259.1 Intron XP_011512561.1
XM_011514261.2 Intron XP_011512563.1
XM_011514262.2 Intron XP_011512564.1
XM_011514263.2 Intron XP_011512565.1
XM_017010198.1 Intron XP_016865687.1
XM_017010199.1 Intron XP_016865688.1
XM_017010200.1 Intron XP_016865689.1
XM_017010201.1 Intron XP_016865690.1
Gene
TTBK1
Gene Name
tau tubulin kinase 1
There are no transcripts associated with this gene.

View Full Product Details