Product Details

SNP ID
rs17038504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:108056528 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGAAAAACTGCAGTCCACGCATAA[C/T]TGAGTCTAACCACACTGTCATTATG
Phenotype
MIM: 153245
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LEF1 PubMed Links

Gene Details

Gene
LEF1
Gene Name
lymphoid enhancer binding factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130713.2 Intron NP_001124185.1
NM_001130714.2 Intron NP_001124186.1
NM_001166119.1 Intron NP_001159591.1
NM_016269.4 Intron NP_057353.1
XM_005263046.3 Intron XP_005263103.1
XM_005263047.1 Intron XP_005263104.1
XM_005263048.1 Intron XP_005263105.1
XM_006714233.1 Intron XP_006714296.1

View Full Product Details