Product Details

SNP ID
rs17130199
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:67703912 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAATCGTTGTAAATAATTACTTC[C/G]TGGGGTTGATTCCTTTTTGAGGGGA
Phenotype
MIM: 615405
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GNG12 PubMed Links

Gene Details

Gene
GNG12
Gene Name
G protein subunit gamma 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018841.5 1963 UTR 3 NP_061329.3
XM_017001809.1 1963 UTR 3 XP_016857298.1
XM_017001810.1 1963 UTR 3 XP_016857299.1
XM_017001811.1 1963 UTR 3 XP_016857300.1

View Full Product Details